Antibody Diluent OP Quanto, Epredia™

Supplier: Epredia
TA-125-ADQ
10129-576EA 190.03 USD
10129-576
Antibody Diluent OP Quanto, Epredia™
Tissue Processors
Achieve crisp staining while minimizing background staining with this staining reagent.

  • Contains a protein stabilizing agent and preservative
  • Concentrated antibodies can be diluted to the appropriate titer specified on the individual antibody datasheet or determined by the user, to achieve optimal staining

Suitable for use as a negative control reagent during immunohistochemical staining.

Storage conditions: 2 - 8 °C
Stability: 24 months

Ordering information: For in vitro diagnostic use.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR