Shandon™ CytoRich™ Red Collection Fluid, Thermo Scientific
Supplier: Epredia
B9990800 B9990801
89404-846EA 260.9 USD
89404-846 89404-848
Shandon™ CytoRich™ Red Collection Fluid, Thermo Scientific
Stains Microscopy and staining reagents
Allow for more accurate evaluation of cytological features with this collection fluid, which lyses RBCs and solubilizes proteins.
  • Pre-filled containers available
  • Ready-to-use, alcohol-based fluid
  • Compatible with immunohistochemistry