Epredia™ Acid-Resistant Biopsy Bags, Epredia
Supplier: Epredia
Epredia™
6774052 6774050 6774051
10170-232CS 287.57 USD
10170-232 10171-092 10171-094
Epredia™ Acid-Resistant Biopsy Bags, Epredia
Bags Sample Bags
Reduce risk of small specimen loss with Epredia™ acid-resistant biopsy bags.
  • Allow unrestricted fluid movement around tissue
  • Reduce reagent carryover and pressure artifact often found with sponges
  • Remove specimen rapidly with convenient peel-open bag
These bags feature all the benefits of nylon biopsy bags, plus offer acid-resistance for decalcification.