Econo™ Sterile Lister Bandage Scissors, Sklar
Supplier: Sklar
96-2502 96-2503 96-2654 96-2500 96-2501
89023-760CS 348.15 USD
89023-760 89023-762 89023-966 75995-372 75995-374
Econo™ Sterile Lister Bandage Scissors, Sklar
Scissors
Econo™ Sterile Lister Bandage Scissors are a versatile instrument with multiple uses.
  • Ready-to-use
  • Angled jaws
  • Lower blade is slightly longer
  • Flattened blunt nodule on tip of lower blade
The scissors are commonly used for sizing dressings and removing circumferential bandages. They may also be used to cut tougher material such as plaster, fabric and occasionally umbilical cord. The jaws of the Lister Bandage Scissors are angled with the lower blade being slightly longer. The tip of the lower blade features a flattened blunt nodule that is intended toMore Product Information