Paraffin wax blended with synthetic polymers, Solidification point: 56-58°C, PureAffin® 1™ for infiltration, without DMSO
Supplier: CANCER DIAGNOSTICS, INC.
PARA1
10810-012CS
511.76
USD
10810-012
Paraffin wax blended with synthetic polymers, Solidification point: 56-58°C, PureAffin® 1™ for infiltration, without DMSO
Paraffin wax blended with synthetic polymers
A purified paraffin specially formulated to provide ideal infiltration with all types of tissue specimens. Tissue Infiltration Medium is compatible with all industry paraffins. Use with open or closed tissue processors. A compound of purified paraffin and additives. Stable at elevated temperatures
- Melting Point 56-58°C (133-136F)
- Working Temperature 60°C (140F)
- Store Below 30°C (86F)
Excellent as two-part paraffin system with PureAffin® 2
CAS Number:
8002-74-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...