VWR® Contour™ ADA Apron Rails
Supplier: VWR International
CFA060336-A CWA060336-A
CFA060336-AEA 180.31 USD
CFA060336-A CWA060336-A
VWR® Contour™ ADA Apron Rails
Furniture Casework
Apron Rails are 3" high (in lieu of standard 4" height) to comply with ADA requirements.
  • Steel
  • Only to be used between base cabinets
A complete apron is used to create a knee space for a desk area or to create a free-standing table (components sold separately).