Adson Suction Tube, OR Grade, Sklar®
Supplier: Sklar
50-1982 50-1985
10199-300EA 150.79 USD
10199-300 10199-302
Adson Suction Tube, OR Grade, Sklar®
Retractors
Sklar's® Adson Suction Tube is used for removal of excess fluids that may be present around a surgical site during a procedure
Removal of these fluids provide the surgeon with a clear view. The Adson Suction Tube is frequently used in neurosurgical or ophthalmologic procedures because of the fine tip that can fit into small sites.
Order Now
SPECIFICATIONS
Environmentally Preferable