ASI Measles, Arlington Scientific
Supplier: ARLINGTON SCIENTIFIC INC.
850096AG 850096AM
10853-082KT 447.03 USD
10853-082 10853-084
ASI Measles, Arlington Scientific
Assays ELISAs
ELISA Kit detects human antibodies to Rubeola Virus.
  • Qualitative and semiquantitative detection
  • Detects human IGG or IGM antibodies
ELISA for the qualitative and semiquantitative detection of human IGG/IGM antibodies to Rubeola Virus.
Packaging: Kit of 96