ASI Ebstein-Barr Viral Capsid Antigen, Arlington Scientific
Supplier: ARLINGTON SCIENTIFIC INC.
880096AG
880096AM
10853-070KT
506.39
USD
10853-070
10853-072
ASI Ebstein-Barr Viral Capsid Antigen, Arlington Scientific
Assays
ELISAs
ELISA Kit detects human antibodies to Ebstein-Barr Viral Capsid Antigen.
- Qualitative and semiquantitative detection
- Detects human IGG or IGM antibodies
ELISA for the qualitative and semiquantitative detection of human IGG/IGM antibodies to Ebstein-Barr viral capsid antigen.
Packaging: Kit of 96