Human Mammary Epithelial Cells, Lifeline Cell Technology
Supplier: Lifeline Cell Technology
FC-0063 FC-0065
102970-716EA 984.77 USD
102970-716 102970-720
Human Mammary Epithelial Cells, Lifeline Cell Technology
Cells
Lifeline Mammary Epithelial Cells are a mixed culture of epithelial cells ensuring optimal morphology and growth.
  • Quality tested
  • Optimal morphology and growth
  • Mixed culture of epithelial cells bearing positive markers
Lifeline® Normal Human Mammary Epithelial Cells are quality tested in MammaryLife™ Medium to ensure optimal morphology and growth over a period of at least 15 population doublings. Lifeline Mammary Epithelial Cells are a mixed culture of epithelial cells bearing positive markers such as Cytokeratin 14 (myoepithelial), Cytokeratin 8/18 (luminal), and Cytokeratin 19 (luminal subpopulation).