Human Prostate Epithelial Cells, Lifeline Cell Technology
Supplier: Lifeline Cell Technology
FC-0038
102970-684EA 1103.69 USD
102970-684
Human Prostate Epithelial Cells, Lifeline Cell Technology
Cells
Lifeline® Normal Human Prostate Epithelial Cells are quality tested to ensure optimal serum-free growth.
  • Quality tested
  • Serum-free growth at rates equal to or greater than serum-supplemented media
  • No exposure to antimicrobials or phenol red
Lifeline® Normal Human Prostate Epithelial Cells are quality tested in Lifeline® ProstaLife™ Medium to ensure optimal serum-free growth over a period of at least 15 population doublings at rates equal to or greater than serum-supplemented media. Lifeline® Human Prostate Epithelial Cells are not exposed to antimicrobials or phenol red when cultured in ProstaLife™ medium.