DermaLife M Neonatal Melanocyte Medium, Lifeline Cell Technology
Supplier: Lifeline Cell Technology
DermaLife M Melanocyte Medium is a low serum, PMA-free, Cholera toxin-free medium optimized for the culture of Human Neonatal Epidermal Melanocytes. DermaLife M contains no phenol red or antimicrobials.
- Contains no phenol red or antimicrobials
- Allows you to prepare fresh medium
- Optimized for the culture of Human Epidermal Melanocytes
DermaLife M Melanocyte Medium is a low serum, PMA-free, Cholera toxin-free medium optimized for the culture of Human Neonatal Epidermal Melanocytes
DermaLife M Melanocyte Neonatal Medium Complete Kit includes: DermaLife Basal Medium - 500 mL and DermaLife M Melanocyte Neonatal LifeFactors®.
DermaLife M Neonatal LifeFactors® Kit includes: rh-Insulin, Ascorbic Acid, L-Glutamine, Epinephrine, StiMel8™, Calcium Chloride, Gentamicin and Amphotericin B. DermaLife Basal Medium requires the appropriate LifeFactors® for appropriate cell culture.
DermaLife M contains no phenol red or antimicrobials.
Ordering information: DermaLife Medium Complete Kits include DermaLife Basal Medium and the associated LifeFactors® Kit. DermaLife Basal Medium requires the appropriate LifeFactors®.