Calcium chloride 0.89M in water

Supplier: Lifeline Cell Technology

Synonyms: Chloro calcium

LS-1062
102970-916EA 67.54 USD
102970-916
Calcium chloride 0.89M in water
Calcium chloride solutions

Calcium Chloride is for use with calcium-free basal medium and human keratinocytes.


  • Used with calcium-free basal medium
  • Used with human keratinocytes


Calcium Chloride (0.89) 1.0 ml is for use with calcium-free Lifeline® DermaLife basal medium and human keratinocytes.

Formula: CaCl₂·2H₂O
MDL Number: MFCD00149613
CAS Number: 10035-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR