Coronoary Artery Smooth Muscle Cells, Lifeline Cell Technology
Supplier: Lifeline Cell Technology
FC-0031
102970-674EA 1103.69 USD
102970-674
Coronoary Artery Smooth Muscle Cells, Lifeline Cell Technology
Cells
Lifeline® Normal Human Coronary Artery Smooth Muscle Cells are quality tested and demonstrate optimal low serum growth.
  • Quality tested in VascuLife® SMC Medium
  • Demonstrate optimal low serum growth
  • No exposure to antimicrobials or phenol red
Lifeline® Normal Human Coronary Artery Smooth Muscle Cells are quality tested in VascuLife® SMC Medium and demonstrate optimal low serum growth over a period of at least 15 population doublings at rates equal to or greater than other serum-supplemented media.
Lifeline® Coronary Artery Smooth Muscle Cells are not exposed to antimicrobials or phenol red when cultured in VascuLife® SMC Medium.