Stemlife MSC-BM Bone Marrow Medium, Lifeline Cell Technology
Supplier: Lifeline Cell Technology
LS-1090 LM-0011 LL-0062
102970-876EA 205.54 USD
102970-876 102970-878 102970-908
Stemlife MSC-BM Bone Marrow Medium, Lifeline Cell Technology
Cell Culture Media
Lifeline® StemLife MSC-BM Bone Marrow Mesenchymal Stem Cells Medium supports stem cell expansion.
  • Undifferentiated expansion of bone marrow mesenchymal stem cells
  • No phenol red
  • No antimicrobials
Lifeline® StemLife MSC-BM Bone Marrow Mesenchymal Stem Cells Medium supports the undifferentiated expansion of bone marrow mesenchymal stem cells. StemLife MSC-BM contains no antimicrobials and no phenol red.
The StemLife MSC-BM Medium Complete Kit includes StemLife MSC-BM Basal Medium and StemLife MSC-BM LifeFactors® Kit. StemLife MSC Basal Medium is designed to be used in conjunction with the associated Lifeline® supplements and growth factors called “LifeFactors®.” StemLife MSC-BM LifeFactors® Kit includes: L-Alanyl-L-Glutamine 6mL, FBS 35 mL, rh IGF-1 0.5 mL, rh FGF b 0.5 mL, Gentamicin and Amphotericin B 0.5 mL.
Ordering information: StemLife MSC-BM Medium Complete Kit includes StemLife MSC Basal Medium and StemLife MSC-BM LifeFactors Kit®. StemLife MSC Basal Medium requires the associated supplements and growth factors.