ViraSafe™ Lentiviral Packaging Systems, Cell Biolabs
Supplier: Cell Biolabs
VPK-205
VPK-206
102966-346EA
892.31
USD
102966-346
102966-348
ViraSafe™ Lentiviral Packaging Systems, Cell Biolabs
Transfection Reagents
ViraSafe™ Lentiviral Packaging Systems utilize 3rd generation lentiviral technology to create recombinant lentivirus when co-transfected along with your 3rd generation lentiviral expression vector
Unlike most commercially available 3rd generation packaging systems, our lentiviral packaging systems are provided as separate plasmids, not a mixture. This allows you to amplify each plasmid individually and eliminates the need to re-order a packaging mixture.