ViraBind™ AAV Purification Kit, Cell Biolabs
Supplier: Cell Biolabs
VPK-140
VPK-141
VPK-141-5
102965-360EA
806.15
USD
102965-360
102965-364
102965-366
ViraBind™ AAV Purification Kit, Cell Biolabs
Nucleic Acid Reagents
Nucleic Acid Purification Kits and Reagents
ViraBind™ AAV Purification Kits use a single-step proprietary purification matrix, followed by further purification and concentration with a centrifugal concentrator
The result is a higher viral yield with exceptional purity in a fraction of the time compared to tedious, time-consuming ultracentrifugation methods. Obtain virus of>95% purity in about 3 hours with >60% recovery.