Ampicillin sodium salt

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: D(-)-α-Aminobenzylpenicillin Sodium Salt

0219014805
IC19014805EA 101.82 USD
IC19014805
Ampicillin sodium salt
Ampicillin sodium salt

Ampicillin is a semi-synthetic derivative of penicillin, active as a broad-spectrum antibiotic.


It is inactivated by beta-lactamases and for this reason a beta-lactamase inhibitor should be considered when using ampicillin. Against gram-positive bacteria, ampicillin has a similar mode of action as benzylpenicillin; against gram-negative bacteria, it has a similar mode of action as chloramphenicol and tetracyclines. In E. coli it inhibits cell wall synthesis.


Used to select for ampicillin resistance in mutated and transformed cells.


A β-lactam antibiotic with an amino group side chain attached to the penicillin structure. Penicillin derivative that inhibits bacterial cell-wall synthesis (peptidoglycan cross-linking) by inactivating transpeptidases on the inner surface of the bacterial cell membrane. Bactericidal only to growing Escherichia coli. Mode of resistance: cleavage of β-lactam ring of ampicillin by β-lactamase. Antimicrobial spectrum: gram-negative and gram-positive bacteria.


Soluble in water (50 mg/ml); soluble in 1 M ammonium hydroxide, dilute acids or bases; practically insoluble in alcohol, chloroform, ether and fixed oils.

Formula: C₁₆H₁₈N₃NaO₄S
MW: 371.39
Storage Temperature: Refrigerator
MDL Number: MFCD00064313
CAS Number: 69-52-3
EINECS: 200-708-1
https://us.vwr.com/stibo/web/std.lang.all/97/19/36419719.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR