Glasgow's Minimum Essential Medium (GMEM) Mammalian Cell Culture Medium, HiMedia
Supplier: HiMedia Laboratories
AT069-5L AT058-1L AT058-20L AT102-5L AT069-5L AT102-1L AT058-5L AT102-5L AT058-1L AT102-1L AT102-20L AT102-20L AT069-20L AT069-1L AT058-20L AT058-5L AT069-1L AT069-20L
95016-490EA 46.13 USD
95016-490 95016-492 CA95016-492 95016-494 CA95016-490 95016-496 CA95016-496 95016-498 CA95016-494 CA95016-488 95016-500 95016-488 CA95016-500 95016-502 CA95016-498 95016-504 CA95016-504 CA95016-502
Glasgow's Minimum Essential Medium (GMEM) Mammalian Cell Culture Medium, HiMedia
Cell Culture Media
Glasgow’s Minimum Essential Medium is a modification of Basal Medium Eagle (BME).
  • Modification of Basal Medium Eagle (BME)
  • Originally used to culture BHK-21 clone 13 cells
  • For investigation of the genetic factors affecting cell competence
Ian Macpherson and Michael Stoker added tryptose phosphate broth and twice the concentration of amino acids and vitamins to BME. The medium was originally used to culture BHK-21 clone 13 cells for investigating the genetic factors affecting cell competence.