TNM-FH Insect Medium, HiMedia
Supplier: HiMedia Laboratories
IM008-1L IM008-10L IM008-20L IM008-5L IM008-5L IML008-5X500ML IM008-20L IM008-1L IM008-50L IML008-2X500ML IM008-50L
95039-080EA 297.07 USD
95039-080 CA95039-080 95038-658 95039-082 95038-656 95037-464 CA95039-078 95037-466 95039-078 CA95038-656 CA95039-082
TNM-FH Insect Medium, HiMedia
Cell Culture Media
TNM-FH medium is a modification of Grace’s insect medium.
  • Modification of Grace’s insect medium
TNM-FH medium is a modification of Grace’s insect medium. The modification was developed by Dr. W. F. Hink to grow Cabbage looper, Tricoplusia.