BootieButler® Waterproof Shoe Covers, Protexer
Supplier: Protexer
KBPE825
10836-055CS 444.68 USD
10836-055
BootieButler® Waterproof Shoe Covers, Protexer
Disposable Garments
Constructed of chlorinated polyethylene (CPE), this waterproof bootie is an ideal low-cost solution for light-duty applications
Strong elastic band around the ankle allows for a universal fit.
Certifications: Approved for Class 100 & Class 5 Cleanroom use per Federal Standard 209E and ISO STD. 14664-1 Class 5.