Ward's® Preserved Dogfish Sharks, 27"+
Supplier: VWR
Ward's®
WARD470001-464 WARD470001-470 WARD470001-758 WARD470001-756 WARD470001-460 WARD470001-760 WARD470001-466
470228-294PK 244.99 USD
470228-294 470228-292 470001-758 470001-470 470001-460 470001-466 470001-760 470001-464 470228-288 470228-290 470001-756
Ward's® Preserved Dogfish Sharks, 27"+
Specimens Preserved Specimens Preserved Animal Materials
Standard formalin preserved dogfish sharks.
  • Cartilaginous fish anatomy
  • No swim bladder or operculum
  • Good for group dissection
  • Gill slits
Dogfish sharks, Squalus species, are a popular teaching specimen for biology and anatomy courses, specifically used for the teaching of cartilaginous fish anatomy. Dogfish will either be species Squalus species or Mustelus canis.
They are also great animal models to study comparative and vertebrate anatomy.
The harvesting of dogfish sharks is regulated by National Marine Fisheries to protect the species. Quantities used by our industry are very small in relation to the overall harvest.
Packaging: Packed in pails or vacuum-sealled bags.