Anti-IgG1 Goat Polyclonal Antibody (IRDye® 800CW)

Supplier: LI-COR®
926-32350
102673-352EA 380.31 USD
102673-352
Anti-IgG1 Goat Polyclonal Antibody (IRDye® 800CW)
Antibodies
Isolation of specific antibodies was accomplished by affinity chromatography using mouse IgG1 covalently linked to agarose beads. Based on ELISA and flow cytometry, this antibody reacts with the heavy chain of mouse IgG1. This antibody was tested by Dot Blot and/or solid phase adsorbed for minimal cross-reactivity with mouse IgM, IgG2a, IgG2b, IgG3, and IgA, pooled human sera and purified human paraproteins. The conjugate has been specifically tested and qualified for Western blot and In-Cell Western™ assay applications. Highly Recommended for: Western blot, In-Cell Western Assay, On-Cell Western Assay, protein array, immunohistochemistry, microsciopy, 2D gel detection, tissue section imaging, virus titration assay, FRET-based assays, and small animal imaging.

Western blot: 1:5000

Type: Secondary
Antigen: IgG1
Clonality: Polyclonal
Clone:
Conjugation: IRDye 800CW
Epitope:
Host: Goat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR