Anti-IgG Goat Polyclonal Antibody (IRDye® 800CW)

Supplier: LI-COR®
926-32210
102673-328EA 249.85 USD
102673-328
Anti-IgG Goat Polyclonal Antibody (IRDye® 800CW)
Antibodies
Isolation of specific antibodies was accomplished by affinity chromatography using pooled mouse IgG covalently linked to agarose beads. Based on ELISA and flow cytometry, this antibody reacts with the heavy and light chains of mouse IgG1, IgG2a, IgG2b, and IgG3, and with the light chains of mouse IgM and IgA. This antibody was tested by Dot Blot and/or solid-phase adsorbed for minimal cross-reactivity with goat, horse, human, rabbit, and rat serum proteins, but may cross-react with immunoglobulins from other species. The conjugate has been specifically tested and qualified for Western blot applications. Highly Recommended for: Western blot, In-Cell Western Assay, On-Cell Western Assay, protein array, immunohistochemistry, microsciopy, 2D gel detection, tissue section imaging, virus titration assay, FRET-based assays, and small animal imaging.

Western blot: 1:5000

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: IRDye 800CW
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR