Anti-IgG2a Goat Polyclonal Antibody (IRDye® 680LT)

Supplier: LI-COR®
926-68051
102673-404EA 380.31 USD
102673-404
Anti-IgG2a Goat Polyclonal Antibody (IRDye® 680LT)
Antibodies
Isolation of specific antibodies was accomplished by affinity chromatography using mouse IgG2a covalently linked to agarose beads. Based on ELISA and flow cytometry, this antibody reacts with the heavy chain of mouse IgG2a. This antibody was tested by Dot Blot and/or solid-phase adsorbed for minimal cross-reactivity with mouse IgM, IgG1, IgG2b, IgG3, IgA, pooled human sera, and purified human paraproteins. The conjugate has been specifically tested and qualified for Western blot applications.

Western blot: 1:20000

Type: Secondary
Antigen: IgG2a
Clonality: Polyclonal
Clone:
Conjugation: IRDye 680LT
Epitope: α chain
Host: Goat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR