Anti-IgG Goat Polyclonal Antibody (IRDye® 680LT)

Supplier: LI-COR®
925-68020
102971-008EA 146.46 USD
102971-008
Anti-IgG Goat Polyclonal Antibody (IRDye® 680LT)
Antibodies
Isolation of specific antibodies was accomplished by affinity chromatography using pooled mouse IgG covalently linked to agarose beads. Based on ELISA and flow cytometry, this antibody reacts with the heavy and light chains of mouse IgG1, IgG2a, IgG2b, and IgG3, and with the light chains of mouse IgM and IgA. This antibody was tested by Dot Blot and/or solid-phase adsorbed for minimal cross-reactivity with human, rabbit, goat, rat, and horse serum proteins, but may cross-react with immunoglobulins from other species. The conjugate has been specifically tested and qualified for Western blot applications. Recommended for: Western blot, proein array, immunohistochemistry, microscopy, 2D gel detection, and tissue section imaging. NOT RECOMMENDED for: small animal imaging, In-Cell Wesern assays, or On-Cell Western assays.

Western blot: 1:20000

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: IRDye 680LT
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR