Anti-IgG Donkey Polyclonal Antibody (IRDye® 680LT)

Supplier: LI-COR®
926-68030
102673-396EA 249.85 USD
102673-396
Anti-IgG Donkey Polyclonal Antibody (IRDye® 680LT)
Antibodies
Isolation of specific antibodies was accomplished by affinity chromatography using antigens covalently linked to agarose beads. Based on immunoelectrophoresis and/or ELISA, this antibody reacts with whole molecule guinea pig IgG as well as the light chains common to other guinea pig immunoglobulins. No reactivity was detected against non-immunoglobulin serum proteins. This antibody was tested by ELISA and/or solid-phase adsorbed for minimal cross-reactivity with bovine, chicken, goat, Syrian hamster, horse, human, mouse, rabbit, rat, and sheep serum proteins, but may cross-react with immunoglobulins from other species. The conjugate has been specifically tested and qualified for Western blot applications. Recommended for: Western blot, proein array, immunohistochemistry, microscopy, 2D gel detection, and tissue section imaging. NOT RECOMMENDED for: small animal imaging, In-Cell Wesern assays, or On-Cell Western assays.

Western blot: 1:20000

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: IRDye 680LT
Epitope:
Host: Donkey
Isotype:
Reactivity: Guinea Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR