CytoSelect™ 24-Well Wound Healing Assay Kit, Cell Biolabs, Inc
Supplier: Cell Biolabs
CBA-120 CBA-120-5 CBA-120-T
102963-766EA 769.23 USD
102963-766 102963-768 102963-770
CytoSelect™ 24-Well Wound Healing Assay Kit, Cell Biolabs, Inc
Assays Immunological Assays
CytoSelect™ 24-Well Wound Healing Assay measures cell migration across a wound field gap in vitro
This assay provides a much more accurate and consistent method compared to the traditional scratch assay. Proprietary inserts generate a consistent 0.9mm wound gap between the cells. Cells may then be treated or monitored for proliferation or migration across the wound field by imaging samples at fixed time points or by time-lapse microscopy. The inserts are inert and leave no residues that could impede proliferation or migration of cells.