Anti-CD186 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: K041E5]

Supplier: Biolegend
356013 356014
10759-422EA 242.2 USD
10759-422 10020-904
Anti-CD186 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: K041E5]
Antibodies
Brilliant Violet 421™ anti-human CD186 (CXCR6) [K041E5]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD186
Clonality: Monoclonal
Clone: K041E5
Conjugation: Brilliant Violet® 421
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR