Anti-Syk Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 4D10.2]

Supplier: Biolegend
644303 644304
10019-856EA 158.7 USD
10019-856 10019-858
Anti-Syk Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 4D10.2]
Antibodies
PE anti-Syk [4D10.2]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: ICFC; Size: 100 tests

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Syk
Clonality: Monoclonal
Clone: 4D10.2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR