Anti-Ki-67 Mouse Monoclonal Antibody (Brilliant Violet® 510) [clone: Ki-67]

Supplier: Biolegend
350517 350518
10759-576EA 242.2 USD
10759-576 10019-448
Anti-Ki-67 Mouse Monoclonal Antibody (Brilliant Violet® 510) [clone: Ki-67]
Antibodies
Brilliant Violet 510™ anti-human Ki-67 [Ki-67]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Bovine; Apps: ICFC, IF; Size: 100 tests

Each lot of this antibody is quality control tested by our Ki-67 staining protocol below. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Ki-67
Clonality: Monoclonal
Clone: Ki-67
Conjugation: Brilliant Violet® 510
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR