Anti-GPR83 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: K07JP05]

Supplier: Biolegend
363305 363306
10021-076EA 172.2 USD
10021-076 10021-078
Anti-GPR83 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: K07JP05]
Antibodies
PE anti-human GPR83 [K07JP05]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: GPR83
Clonality: Monoclonal
Clone: K07JP05
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR