Anti-CD8 Mouse Monoclonal Antibody (Brilliant Violet® 510) [clone: SK1]

Supplier: Biolegend
344731 344732
10019-434EA 154.3 USD
10019-434 10019-436
Anti-CD8 Mouse Monoclonal Antibody (Brilliant Violet® 510) [clone: SK1]
Antibodies
Brilliant Violet 510™ anti-human CD8 [SK1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: African Green, Chimpanzee, Cynomolgus, Pigtailed Macaque, Rhesus, Sooty Mangabey; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD8
Clonality: Monoclonal
Clone: SK1
Conjugation: Brilliant Violet® 510
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: African Green;Chimpanzee;Cynomolgus;Pigtailed Macaque;Rhesus;Sooty Mangabey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR