Anti-CD62P (P-selectin) Mouse Monoclonal Antibody [clone: RMP-1]

Supplier: Biolegend
148301 148302
10019-146EA 119.4 USD
10019-146 10019-148
Anti-CD62P (P-selectin) Mouse Monoclonal Antibody [clone: RMP-1]
Antibodies
Purified anti-mouse/rat CD62P (P-selectin) [RMP-1]; Isotype: Mouse IgG2a, κ; Reactivity: Mouse, Rat; Apps: FC, IF, IHC, ELISA, WB, IP; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD62P (P-selectin)
Clonality: Monoclonal
Clone: RMP-1
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR