Anti-CD177 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: MEM-166]

Supplier: Biolegend
315807 315808
10019-264EA 151 USD
10019-264 10020-394
Anti-CD177 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: MEM-166]
Antibodies
APC anti-human CD177 [MEM-166]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD177
Clonality: Monoclonal
Clone: MEM-166
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR