Anti-CD14 Mouse Monoclonal Antibody [clone: 63D3]

Supplier: Biolegend
367101 367102
10019-636EA 60.8 USD
10019-636 10019-638
Anti-CD14 Mouse Monoclonal Antibody [clone: 63D3]
Antibodies
Purified anti-human CD14 [63D3]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 2.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: 63D3
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR