Anti-Tubulin Beta 3 Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: AA10]

Supplier: Biolegend
657405 657406
10763-570EA 198.1 USD
10763-570 10763-572
Anti-Tubulin Beta 3 Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: AA10]
Antibodies
Alexa Fluor® 647 anti-Tubulin Beta 3 (TUBB3) [AA10]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Mouse, Rat; Apps: ICFC, IHC-F; Size: 100 μg

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.06 μg per million cells in 100 μl volume. For immunohistochemical staining on frozen tissue sections, the suggested use is 1.25-5 μg/mL. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Tubulin Beta 3
Clonality: Monoclonal
Clone: AA10
Conjugation: Alexa Fluor® 647
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR