Anti-LEF1 Mouse Monoclonal Antibody [clone: 15H5A18]

Supplier: Biolegend
653101 653102
10763-370EA 113.7 USD
10763-370 10763-372
Anti-LEF1 Mouse Monoclonal Antibody [clone: 15H5A18]
Antibodies
Purified anti-LEF1 [15H5A18]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: WB, IP; Size: 100 μg

Each lot of this antibody is quality control tested by Western blotting. For Western blotting, the suggested use of this reagent is 0.1-0.5 μg per ml. For immunoprecipitation, the suggested use of this reagent is 5-20 μg per ml. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: LEF1
Clonality: Monoclonal
Clone: 15H5A18
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR