Anti-IL-33Ralpha Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: DIH9]

Supplier: Biolegend
145303 145304
10756-456EA 164.4 USD
10756-456 10756-458
Anti-IL-33Ralpha Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: DIH9]
Antibodies
PE anti-mouse IL-33Rα (IL1RL1, ST2) [DIH9]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IL-33Ralpha
Clonality: Monoclonal
Clone: DIH9
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: Rat IgG2a, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR