Anti-IgG2b, k Isotype Ctrl Mouse Monoclonal Antibody (Brilliant Violet® 650) [clone: MPC-11]

Supplier: Biolegend
400351 400352
10761-126EA 240.22 USD
10761-126 10761-128
Anti-IgG2b, k Isotype Ctrl Mouse Monoclonal Antibody (Brilliant Violet® 650) [clone: MPC-11]
Antibodies
Brilliant Violet 650™ Mouse IgG2b, κ Isotype Ctrl [MPC-11]; Isotype: Mouse IgG2b, κ; Reactivity: Mouse; Apps: FC, ICFC; Size: 100 tests

Each lot of this mouse IgG2b, κ isotype control antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, use the isotype control at the same concentration as your primary antibody. Use our Concentration Lookup tool to find the exact concentrations of your lots of product.

Type: Control
Antigen: IgG2b, k Isotype Ctrl
Clonality: Monoclonal
Clone: MPC-11
Conjugation: Brilliant Violet® 650
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR