Anti-HMGB1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 3E8]

Supplier: Biolegend
651403 651404
10763-410EA 158.7 USD
10763-410 10763-412
Anti-HMGB1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 3E8]
Antibodies
PE anti-HMGB1 [3E8]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Mouse, Rat; Apps: ICFC; Size: 100 tests

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining using our nuclear factor staining protocol. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: HMGB1
Clonality: Monoclonal
Clone: 3E8
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR