Anti-H-2Kb bound to SIINFEKL Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 25-D1.16]

Supplier: Biolegend
141605 141606
141605-BLEA 161.8 USD
141605-BL 141606-BL
Anti-H-2Kb bound to SIINFEKL Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 25-D1.16]
Antibodies
APC anti-mouse H-2Kb bound to SIINFEKL [25-D1.16]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: FC, IF; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: H-2Kb bound to SIINFEKL
Clonality: Monoclonal
Clone: 25-D1.16
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR