Anti-GITR Rat Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®) [clone: DTA-1]

Supplier: Biolegend
126315 126316
10757-048EA 165.5 USD
10757-048 10757-050
Anti-GITR Rat Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®) [clone: DTA-1]
Antibodies
PerCP/Cy5.5 anti-mouse CD357 (GITR) [DTA-1]; Isotype: Rat IgG2b, λ; Reactivity: mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: GITR
Clonality: Monoclonal
Clone: DTA-1
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
Epitope:
Host: Rat
Isotype: Rat IgG2b, λ
Reactivity: mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR