Anti-DR3 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: 4C12]

Supplier: Biolegend
144405 144406
10757-352EA 185.7 USD
10757-352 10757-354
Anti-DR3 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: 4C12]
Antibodies
PE anti-mouse DR3 (TNFRSF25) [4C12]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: DR3
Clonality: Monoclonal
Clone: 4C12
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR