Anti-CXCL5 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: J111B7]

Supplier: Biolegend
524103 524104
10762-006EA 172.2 USD
10762-006 10762-152
Anti-CXCL5 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: J111B7]
Antibodies
PE anti-human CXCL5 [J111B7]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: ICFC; Size: 100 tests

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CXCL5
Clonality: Monoclonal
Clone: J111B7
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR