Anti-CD90.2 Rat Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 53-2.1]

Supplier: Biolegend
140315 140316
140315-BLEA 119.4 USD
140315-BL 140316-BL
Anti-CD90.2 Rat Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 53-2.1]
Antibodies
PerCP anti-mouse CD90.2 (Thy1.2) [53-2.1]; Isotype: Rat IgG2a, κ; Reactivity: Mouse, Does not react with Thy-1.1 (CD90.1); Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.06 μg per million cells in 100 μl volume.  It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD90.2
Clonality: Monoclonal
Clone: 53-2.1
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse;Does not react with Thy-1.1 (CD90.1)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR