Anti-CD90.2 Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 53-2.1]

Supplier: Biolegend
140309 140310
140309-BLEA 113.7 USD
140309-BL 140310-BL
Anti-CD90.2 Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 53-2.1]
Antibodies
PE/Cy7 anti-mouse CD90.2 (Thy1.2) [53-2.1]; Isotype: Rat IgG2a, κ; Reactivity: Mouse, Does not react with Thy-1.1 (CD90.1); Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.06 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD90.2
Clonality: Monoclonal
Clone: 53-2.1
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse;Does not react with Thy-1.1 (CD90.1)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR