Anti-CD8b Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: YTS156.7.7]

Supplier: Biolegend
126617 126618
10757-066EA 139.6 USD
10757-066 10757-068
Anti-CD8b Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: YTS156.7.7]
Antibodies
Alexa Fluor® 700 anti-mouse CD8b (Ly-3) [YTS156.7.7]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD8b
Clonality: Monoclonal
Clone: YTS156.7.7
Conjugation: Alexa Fluor® 700
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR