Anti-CD8b Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: YTS156.7.7]

Supplier: Biolegend
126613 126614
10757-058EA 120.03 USD
10757-058 10757-060
Anti-CD8b Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: YTS156.7.7]
Antibodies
APC anti-mouse CD8b (Ly-3) [YTS156.7.7]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.125 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD8b
Clonality: Monoclonal
Clone: YTS156.7.7
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR