Anti-CD8a Rat Monoclonal Antibody (PE (Phycoerythrin)/Dazzle 594) [clone: 53-6.7]

Supplier: BioLegend
100762 100762 100761 100761
10756-714EA 417.71 USD
10756-714 CA10756-712 10756-712 CA10756-714
Anti-CD8a Rat Monoclonal Antibody (PE (Phycoerythrin)/Dazzle 594) [clone: 53-6.7]
Antibodies
PE/Dazzle™ 594 anti-mouse CD8a [53-6.7]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.125 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD8a
Clonality: Monoclonal
Clone: 53-6.7
Conjugation: PE (Phycoerythrin)/Dazzle 594
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR